missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, HPLC Grade, 99.7+%

Catalog No. p-4782010
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L
6 x 1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA22915K2 1 L
AA22915M1 4 x 1 L
AA22915K7 4 L
AA22915Y0 6 x 1 L
AA22915M6 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AA22915K2 Supplier Thermo Scientific Chemicals Supplier No. 022915K2
Only null left

CAS: 68-12-2 | C3H7NO | 73.10 g/mol

N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films. It is used as a mobile phase in High Performance Liquid Chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Melting Point 61°C
Color Colorless
Appearance Colorless liquid
Density 0.944 g/cm3 at 20°C
Quantity 1 L
Flash Point 57°C (135°F)
Odor Amine-like
Physical Form Liquid
Refractive Index 1.431
Assay Percent Range >99.7%
Vapor Pressure 3.5 hPa (3mm Hg) at 20°C
UN Number 2265
Beilstein 605365
Boiling Point 153°C
Solubility Information Miscible with water and most organic solvents.
Formula Weight 73.09
Grade HPLC
Chemical Name or Material N, N-Dimethylformamide
Show More Show Less
Hazard Category H226-H312+H332-H319-H335-H336-H350-H360D
Hazard Statement Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P312-P363-P370+P378q-P501c
DOTInformation DOT Class: 3; Packing Group: III
EINECSNumber 200-679-5
RTECSNumber LQ2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only