missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, For Spectroscopy Uvasol™, MilliporeSigma™

Catalog No. M1029370500
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1029370500 500 mL Glass Bottle
M1029372500 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1029370500 Supplier Technidata Supplier No. 1.02937.0500

For spectroscopy Uvasol™

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

pH 7 (at 200g/L 20°C)
Melting Point -61°C
Color Colorless
Density 0.94g/cm3 at 20°C
Quantity 500 mL
Flash Point 58°C (136°F)
Ignition Point 410°C (770°F)
Odor Weakly amine-like
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.43
Vapor Pressure 3.77 hPa (20°C)
UN Number 2265
Boiling Point 153°C
Solubility Information Soluble in water at 20°C
Formula Weight 73.09
Chemical Name or Material N,N-Dimethylformamide
Show More Show Less
Hazard Category H360D May damage the unborn child.
Hazard Statement H226 Flammable liquid and vapor.
Precautionary Statement H312 + H332 Harmful in contact with skin or if inhaled.
Recommended Storage 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition. Keep locked up or in an area accessible only to qualified or authorized persons.