missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylformamide Dimethyl Acetal 96.0+%, TCI America™
Supplier: TCI America D1293500ML
Specifications
| N,N-Dimethylformamide Dimethyl Acetal | |
| Yellow | |
| C5H13NO2 | |
| 500 mL | |
| n,n-dimethylformamide dimethyl acetal, 1,1-dimethoxytrimethylamine, dimethoxymethyl dimethylamine, methanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylformamide-dimethylacetal, dmf dimethyl acetal, dmfdma, dimethylformamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylmethylamine, dimethoxy dimethylamino methane | |
| COC(OC)N(C)C | |
| 119.16 | |
| CHEBI:85061 | |
| ≥96.0% (GC,T) |
| 4637-24-5 | |
| 104°C | |
| MFCD00008482 | |
| 1993 | |
| ZSXGLVDWWRXATF-UHFFFAOYSA-N | |
| (dimethoxymethyl)dimethylamine | |
| 78373 | |
| 119.16 | |
| Liquid |
Chemical Identifiers
| 4637-24-5 | |
| 119.16 | |
| ZSXGLVDWWRXATF-UHFFFAOYSA-N | |
| 78373 | |
| (dimethoxymethyl)dimethylamine |
| C5H13NO2 | |
| MFCD00008482 | |
| n,n-dimethylformamide dimethyl acetal, 1,1-dimethoxytrimethylamine, dimethoxymethyl dimethylamine, methanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylformamide-dimethylacetal, dmf dimethyl acetal, dmfdma, dimethylformamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylmethylamine, dimethoxy dimethylamino methane | |
| CHEBI:85061 | |
| COC(OC)N(C)C |
Safety and Handling
EINECSNumber : (2)-3547
TSCA : Yes