missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide Dimethyl Acetal 96.0+%, TCI America™

Catalog No. D129325ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D129325ML 25 mL
D1293100ML 100 mL
D1293500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D129325ML Supplier TCI America Supplier No. D129325ML

Chemical Identifiers

CAS 4637-24-5
Molecular Formula C5H13NO2
Molecular Weight (g/mol) 119.16
MDL Number MFCD00008482
InChI Key ZSXGLVDWWRXATF-UHFFFAOYSA-N
Synonym n,n-dimethylformamide dimethyl acetal, 1,1-dimethoxytrimethylamine, dimethoxymethyl dimethylamine, methanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylformamide-dimethylacetal, dmf dimethyl acetal, dmfdma, dimethylformamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylmethylamine, dimethoxy dimethylamino methane
PubChem CID 78373
ChEBI CHEBI:85061
IUPAC Name (dimethoxymethyl)dimethylamine
SMILES COC(OC)N(C)C

Specifications

Color Yellow
Boiling Point 104°C
Quantity 25 mL
UN Number 1993
Formula Weight 119.16
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Dimethylformamide Dimethyl Acetal
EINECSNumber (2)-3547
TSCA Yes