missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide dimethyl acetal, ca. 97%, AcroSeal™

Catalog No. AC446151000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC446151000 100 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC446151000 Supplier Thermo Scientific Chemicals Supplier No. 446151000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4637-24-5 | C5H13NO2 | 119.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent

Chemical Identifiers

CAS 4637-24-5 , 149-73-5 , 68-12-2 , 67-56-1
Molecular Formula C5H13NO2
Molecular Weight (g/mol) 119.16
MDL Number MFCD00008482
InChI Key ZSXGLVDWWRXATF-UHFFFAOYSA-N
Synonym n,n-dimethylformamide dimethyl acetal, 1,1-dimethoxytrimethylamine, dimethoxymethyl dimethylamine, methanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylformamide-dimethylacetal, dmf dimethyl acetal, dmfdma, dimethylformamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylmethylamine, dimethoxy dimethylamino methane
PubChem CID 78373
ChEBI CHEBI:85061
IUPAC Name (dimethoxymethyl)dimethylamine
SMILES COC(OC)N(C)C

Specifications

Color White
Density 0.8900g/mL
Boiling Point 102.0°C to 104.0°C
Flash Point 7°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging AcroSeal™ Glass bottle
Refractive Index 1.3972
Linear Formula (CH3)2NCH(OCH3)2
Quantity 100 mL
Specific Gravity 0.89
Solubility Information Solubility in water: hydrolysis. Other solubilities: soluble in many organic solvents
Formula Weight 119.16
Percent Purity ca. 97%
Physical Form Solid
Chemical Name or Material N, N-Dimethylformamide dimethyl acetal
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye damage.
May cause an allergic skin reaction.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF exposed or concerned: Get medical attention/advice.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face p
EINECSNumber 225-063-3

RUO – Research Use Only