Learn More
N,N-Dimethylformamide dimethyl acetal, 94%
CAS: 4637-24-5 | C5H13NO2 | 119.16 g/mol
Supplier: Thermo Scientific Chemicals A1535022
Description
N,N-Dimethylformamide dimethyl acetal is used as an intermediate in the formation of pyridine derivatives, which exhibits inhibition against PI3 kinase p110alfa enzymes. It is utilized for the derivatization of primary sulfonamides and trifluoroacetic acid. It is also used in the preparation of formamidine derivatives. It is used as a reagent for n-dimethylaminomethylene and methyl esters. Further, it is used to catalyze the coupling of epoxides with carbon dioxide to prepare cyclic carbonates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N-Dimethylformamide dimethyl acetal is used as an intermediate in the formation of pyridine derivatives, which exhibits inhibition against PI3 kinase p110alfa enzymes. It is utilized for the derivatization of primary sulfonamides and trifluoroacetic acid. It is also used in the preparation of formamidine derivatives. It is used as a reagent for n-dimethylaminomethylene and methyl esters. Further, it is used to catalyze the coupling of epoxides with carbon dioxide to prepare cyclic carbonates.
Solubility
Miscible with most organic solvents.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents and acids.
Specifications
| N,N-Dimethylformamide dimethyl acetal | |
| -85°C | |
| 102°C to 104°C | |
| C5H13NO2 | |
| MFCD00008482 | |
| UN1993 | |
| n,n-dimethylformamide dimethyl acetal, 1,1-dimethoxytrimethylamine, dimethoxymethyl dimethylamine, methanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylformamide-dimethylacetal, dmf dimethyl acetal, dmfdma, dimethylformamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylmethylamine, dimethoxy dimethylamino methane | |
| ZSXGLVDWWRXATF-UHFFFAOYSA-N | |
| (dimethoxymethyl)dimethylamine | |
| 78373 | |
| 119.16 |
| 4637-24-5 | |
| 0.897 | |
| 7°C (45°F) | |
| 1.3972 | |
| 100 g | |
| Moisture sensitive | |
| Miscible with most organic solvents. | |
| COC(OC)N(C)C | |
| 119.16 | |
| CHEBI:85061 | |
| 97% |
Chemical Identifiers
| 4637-24-5 | |
| 119.16 | |
| ZSXGLVDWWRXATF-UHFFFAOYSA-N | |
| 78373 | |
| (dimethoxymethyl)dimethylamine |
| C5H13NO2 | |
| MFCD00008482 | |
| n,n-dimethylformamide dimethyl acetal, 1,1-dimethoxytrimethylamine, dimethoxymethyl dimethylamine, methanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylformamide-dimethylacetal, dmf dimethyl acetal, dmfdma, dimethylformamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylmethylamine, dimethoxy dimethylamino methane | |
| CHEBI:85061 | |
| COC(OC)N(C)C |
Safety and Handling
GHS H Statement
H225-H302-H332-H315-H319-H335
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P210-P233-P240-P241-P242-P243-P261-P271-P272-P280g-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P312-P333+P313-P363-P370+P378q-P372-P374-P380-P501c
H225-H317-H318-H332-H361
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 225-063-3
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only