missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide dimethyl acetal, >95%, MilliporeSigma™ Supelco™

Catalog No. p-7187686
Change view
Click to view available options
Quantity:
5 mL
25 mL
10 x 1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019201 5 mL
111018746 10 x 1 mL
111018564 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111019201 Supplier Merck Emd Millipore Supplier No. 3949635ML
Only null left

Esterate M (N,N-Dimethylformamide dimethyl acetal, DMFDMA) is an methylating reagent. It has been used in methyl esterification of carboxylic acid and also in one-step derivatization of amino acids into N,N-Dimethylaminomethylene methyl esters.

Chemical Identifiers

MDL Number MFCD00008482
Synonym 1,1-Dimethoxy-N,N-dimethylmethylamine; 1,1-Dimethoxytrimethylamine

Specifications

Density 0.897 g/mL (at 25°C)
CAS 4637-24-5
Boiling Point 102°C to 103°C (720 mmHg)
Flash Point 5°C - Closed Cup
Packaging Ampule
Linear Formula (CH3)2 NCH(OCH3)2
Quantity 5 mL
Formula Weight 119.16
Percent Purity >95%
Grade For GC derivatization