missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide Diethyl Acetal 95.0+%, TCI America™

Catalog No. p-7126663
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12945ML 5 mL
D129425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D12945ML Supplier TCI America Supplier No. D12945ML
Only null left

Chemical Identifiers

CAS 1188-33-6
Molecular Formula C7H17NO2
Molecular Weight (g/mol) 147.218
MDL Number MFCD00009227
InChI Key BWKAYBPLDRWMCJ-UHFFFAOYSA-N
Synonym n,n-dimethylformamide diethyl acetal, 1,1-diethoxytrimethylamine, methanamine, 1,1-diethoxy-n,n-dimethyl, n,n-dimethyformamide diethy acetal, diethoxymethyl dimethylamine, n,n-dimethylformamide diethylacetal, dimethylformamide diethylacetal, 1,1-diethoxy trimethyl amine, 1,1-diethoxy-n,n-dimethylmethylamine, 1,1-diethoxy-n,n-dimethyl-methanamine
PubChem CID 70913
IUPAC Name 1,1-diethoxy-N,N-dimethylmethanamine
SMILES CCOC(N(C)C)OCC

Specifications

Color Yellow
Boiling Point 130°C
Quantity 5 mL
UN Number 1993
Formula Weight 147.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Dimethylformamide Diethyl Acetal [for Esterification]