missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide diethyl acetal, 95%

CAS: 1188-33-6 | C7H17NO2 | 147.22 g/mol

$288.45 - $288.45

Chemical Identifiers

CAS 1188-33-6
Molecular Formula C7H17NO2
Molecular Weight (g/mol) 147.22
MDL Number MFCD00009227
InChI Key BWKAYBPLDRWMCJ-UHFFFAOYSA-N
Synonym n,n-dimethylformamide diethyl acetal, 1,1-diethoxytrimethylamine, methanamine, 1,1-diethoxy-n,n-dimethyl, n,n-dimethyformamide diethy acetal, diethoxymethyl dimethylamine, n,n-dimethylformamide diethylacetal, dimethylformamide diethylacetal, 1,1-diethoxy trimethyl amine, 1,1-diethoxy-n,n-dimethylmethylamine, 1,1-diethoxy-n,n-dimethyl-methanamine
PubChem CID 70913
IUPAC Name 1,1-diethoxy-N,N-dimethylmethanamine
SMILES CCOC(N(C)C)OCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC162080500
SDS View Documents
Thermo Scientific Chemicals
162080500
50 g
Each for $288.45
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
Specifications

Chemical Identifiers

1188-33-6
147.22
BWKAYBPLDRWMCJ-UHFFFAOYSA-N
70913
CCOC(N(C)C)OCC
C7H17NO2
MFCD00009227
n,n-dimethylformamide diethyl acetal, 1,1-diethoxytrimethylamine, methanamine, 1,1-diethoxy-n,n-dimethyl, n,n-dimethyformamide diethy acetal, diethoxymethyl dimethylamine, n,n-dimethylformamide diethylacetal, dimethylformamide diethylacetal, 1,1-diethoxy trimethyl amine, 1,1-diethoxy-n,n-dimethylmethylamine, 1,1-diethoxy-n,n-dimethyl-methanamine
1,1-diethoxy-N,N-dimethylmethanamine

Specifications

1188-33-6
100.0
128°C to 130°C
94% min. (GC)
C7H17NO2
MFCD00009227
01,281; 02,154; 05,253; 07,125; 10,157
n,n-dimethylformamide diethyl acetal, 1,1-diethoxytrimethylamine, methanamine, 1,1-diethoxy-n,n-dimethyl, n,n-dimethyformamide diethy acetal, diethoxymethyl dimethylamine, n,n-dimethylformamide diethylacetal, dimethylformamide diethylacetal, 1,1-diethoxy trimethyl amine, 1,1-diethoxy-n,n-dimethylmethylamine, 1,1-diethoxy-n,n-dimethyl-methanamine
BWKAYBPLDRWMCJ-UHFFFAOYSA-N
1,1-diethoxy-N,N-dimethylmethanamine
70913
95%
94.0
0.8590g/mL
28°C
Glass bottle
(CH3)2NCH(OC2H5)2
50 g
0.859
Solubility in water: decomposes
CCOC(N(C)C)OCC
147.22
147.22
N,N-Dimethylformamide diethyl acetal
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

GHS Signal Word: Warning

EINECSNumber : 214-707-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only