missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide diethyl acetal, 95%

Catalog No. p-7058251
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0020509 10 g
AAL0020518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0020509 Supplier Thermo Scientific Chemicals Supplier No. L0020509
Only null left

CAS: 1188-33-6 | C7H17NO2 | 147.218 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1188-33-6
Molecular Formula C7H17NO2
Molecular Weight (g/mol) 147.218
MDL Number MFCD00009227
InChI Key BWKAYBPLDRWMCJ-UHFFFAOYSA-N
Synonym n,n-dimethylformamide diethyl acetal, 1,1-diethoxytrimethylamine, methanamine, 1,1-diethoxy-n,n-dimethyl, n,n-dimethyformamide diethy acetal, diethoxymethyl dimethylamine, n,n-dimethylformamide diethylacetal, dimethylformamide diethylacetal, 1,1-diethoxy trimethyl amine, 1,1-diethoxy-n,n-dimethylmethylamine, 1,1-diethoxy-n,n-dimethyl-methanamine
PubChem CID 70913
IUPAC Name 1,1-diethoxy-N,N-dimethylmethanamine
SMILES CCOC(N(C)C)OCC

Specifications

Density 0.859
Boiling Point 134°C to 136°C
Flash Point 17°C (62°F)
Refractive Index 1.399
Quantity 10 g
UN Number UN1993
Beilstein 741889
Sensitivity Moisture sensitive
Formula Weight 147.22
Percent Purity 95%
Chemical Name or Material N,N-Dimethylformamide diethyl acetal
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 214-707-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only