missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide Dibutyl Acetal 98.0+%, TCI America™

Catalog No. D13025ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D13025ML 5 mL
D130225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D13025ML Supplier TCI America Supplier No. D13025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18503-90-7
Molecular Formula C11H25NO2
Molecular Weight (g/mol) 203.33
MDL Number MFCD00015249
InChI Key GSFFXKGTGPMVLM-UHFFFAOYSA-N
Synonym 1,1-Dibutoxytrimethylamine
PubChem CID 87683
IUPAC Name (dibutoxymethyl)dimethylamine
SMILES CCCCOC(OCCCC)N(C)C

Specifications

Color Yellow
Boiling Point 98°C
Quantity 5 mL
UN Number 1993
Formula Weight 203.33
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Dimethylformamide Dibutyl Acetal [for Esterification]