missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide di-tert-butyl Acetal, tech. 90%

Catalog No. AAL0043106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0043106 5 g
AAL0043114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0043106 Supplier Thermo Scientific Chemicals Supplier No. L0043106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 36805-97-7 | C11H25NO2 | 203.326 g/mol

N,N-Dimethylformamide di-tert-butyl acetal is used as a reagent in the synthesis of (S)-2-(1-tert-butoxycarbonyl-2-{4-[2-(5-methyl-2-phenyloxazol-4-yl)ethoxy]phenyl}ethylamino)benzoic acid methyl ester, 3-O-tert-butylmorphine, tert-butylesters of pyrrole and indolecarboxylic acids. Further, it is used in the preparation of indolizines by intermolecular cyclization of picolinium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylformamide di-tert-butyl acetal is used as a reagent in the synthesis of (S)-2-(1-tert-butoxycarbonyl-2-{4-[2-(5-methyl-2-phenyloxazol-4-yl)ethoxy]phenyl}ethylamino)benzoic acid methyl ester, 3-O-tert-butylmorphine, tert-butylesters of pyrrole and indolecarboxylic acids. Further, it is used in the preparation of indolizines by intermolecular cyclization of picolinium salts.

Solubility
Miscible with toluene and benzene.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and acids.

Chemical Identifiers

CAS 36805-97-7
Molecular Formula C11H25NO2
Molecular Weight (g/mol) 203.326
MDL Number MFCD00015002
InChI Key DBNQIOANXZVWIP-UHFFFAOYSA-N
Synonym n,n-dimethylformamide di-tert-butyl acetal, 1,1-di-tert-butoxytrimethylamine, 1,1-di-tert-butoxy-n,n-dimethylmethanamine, bis tert-butoxy methyl dimethylamine, 1,1-bis 1,1-dimethylethoxy-n,n,n-trimethylamine, di-tert-butoxymethyldimethylamine, n,n-dimethylformamide di-t-butyl acetal, methanamine, 1,1-bis 1,1-dimethylethoxy-n,n-dimethyl, 1,1-ditert-butoxy-n,n-dimethylmethanamine, 1,1-di-tert-butoxy-n,n-dimethylmethylamine
PubChem CID 547712
IUPAC Name N,N-dimethyl-1,1-bis[(2-methylpropan-2-yl)oxy]methanamine
SMILES CC(C)(C)OC(N(C)C)OC(C)(C)C

Specifications

Density 0.85
Boiling Point 75°C to 77°C (12 mmHg)
Flash Point 33°C (91°F)
Refractive Index 1.412
Linear Formula (CH3)2NCH[OC(CH3)3]2
Quantity 5 g
UN Number UN1993
Beilstein 969629
Sensitivity Moisture sensitive
Solubility Information Miscible with toluene and benzene.
Formula Weight 203.33
Percent Purity 90%
Grade Technical
Chemical Name or Material N,N-Dimethylformamide di-tert-butyl acetal
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 253-222-7
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only