missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide Di-tert-butyl Acetal 98.0+%, TCI America™

Catalog No. p-7126668
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D13035ML 5 mL
D130325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D13035ML Supplier TCI America Supplier No. D13035ML
Only null left

Chemical Identifiers

CAS 36805-97-7
Molecular Formula C11H25NO2
Molecular Weight (g/mol) 203.326
MDL Number MFCD00015002
InChI Key DBNQIOANXZVWIP-UHFFFAOYSA-N
Synonym n,n-dimethylformamide di-tert-butyl acetal, 1,1-di-tert-butoxytrimethylamine, 1,1-di-tert-butoxy-n,n-dimethylmethanamine, bis tert-butoxy methyl dimethylamine, 1,1-bis 1,1-dimethylethoxy-n,n,n-trimethylamine, di-tert-butoxymethyldimethylamine, n,n-dimethylformamide di-t-butyl acetal, methanamine, 1,1-bis 1,1-dimethylethoxy-n,n-dimethyl, 1,1-ditert-butoxy-n,n-dimethylmethanamine, 1,1-di-tert-butoxy-n,n-dimethylmethylamine
PubChem CID 547712
IUPAC Name N,N-dimethyl-1,1-bis[(2-methylpropan-2-yl)oxy]methanamine
SMILES CC(C)(C)OC(N(C)C)OC(C)(C)C

Specifications

Color Colorless
Boiling Point 57°C
Quantity 5 mL
UN Number 1993
Formula Weight 203.33
Percent Purity ≥98.0% (N)
Physical Form Liquid
Chemical Name or Material N,N-Dimethylformamide Di-tert-butyl Acetal [for Esterification]