missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylformamide Di-tert-butyl Acetal 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 57°C |
| Quantity | 25 mL |
| UN Number | 1993 |
| Formula Weight | 203.33 |
| Percent Purity | ≥98.0% (N) |
| Physical Form | Liquid |
| Chemical Name or Material | N,N-Dimethylformamide Di-tert-butyl Acetal [for Esterification] |
Safety and Handling
| TSCA | No |