missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide di-n-propyl acetal, 97%

Catalog No. AAL0064309
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0064309 10 g
AAL0064318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0064309 Supplier Thermo Scientific Chemicals Supplier No. L0064309
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6006-65-1 | C9H22NO2 | 176.28 g/mol

N,N-Dimethylformamide di-n-propyl acetal, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylformamide di-n-propyl acetal, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
It hydrolyzes with water.

Notes
Moisture, water Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 6006-65-1
Molecular Formula C9H22NO2
Molecular Weight (g/mol) 176.28
MDL Number MFCD00009374
InChI Key NSLGQFIDCADTAS-UHFFFAOYSA-O
Synonym n,n-dimethylformamide dipropyl acetal, 1,1-dipropoxytrimethylamine, n,n-dimethylformamide di-n-propyl acetal, dipropoxymethyl dimethylamine, 1,1-dipropoxy-n,n-dimethylmethylamine, acmc-1b5t3, dimethyl dipropoxymethyl amine, dimethylformamide dipropyl acetal, nslgqfidcadtas-uhfffaoysa, n,n-dimethylformamide dipropylacetal
PubChem CID 80105
IUPAC Name N,N-dimethyl-1,1-dipropoxymethanamine
SMILES CCCOC(OCCC)[NH+](C)C

Specifications

Density 0.854
Boiling Point 67°C to 68°C (14 mmHg)
Flash Point 46°C (115°F)
Refractive Index 1.4088
Linear Formula (CH3)2NCH(OCH2CH2CH3)2
Quantity 10 g
UN Number UN1993
Beilstein 1098524
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes with water.
Formula Weight 175.27
Percent Purity 97%
Chemical Name or Material N,N-Dimethylformamide di-n-propyl acetal
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 227-855-4
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only