missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide CHROMASOLV™ Plus, for HPLC, ≥99.9%, Solstice

Catalog No. 6000998
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000998 2.5 L
6000997 1 L
6001000 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6000998 Supplier Solstice Supplier No. 2705472.5L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Solvent for many hydrophobic organic compounds.

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Melting Point -61°C
Density 0.944g/mL
Quantity 2.5 L
Refractive Index n20/D 1.430
Linear Formula HCON(CH3)2
Vapor Pressure 2.7mmHg (20 °C)
Beilstein 605365
Boiling Point 153°C
Formula Weight 73.09g/mol
Percent Purity ≥99.9%
Grade glass distillation
Chemical Name or Material N,N-Dimethylformamide, for HPLC
Show More Show Less
Hazard Statement P201-P280-P305 + P351 + P338-P308 + P313
Precautionary Statement H226-H312 + H332-H319-H360D
EINECSNumber 200-679-5
RTECSNumber LQ2100000