missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide (Certified ACS), Fisher Chemical™

Catalog No. p-215895
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Glass Bottle
HDPE Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
D1191 1 L Glass Bottle
D1194 4 L Glass Bottle
D11920 20 L Poly Pail
D119200 200 L HDPE Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. D1191 Supplier Thermo Fisher Scientific Supplier No. BPD1191
Edge

Meet or exceeds the latest ACS specifications. Actual lot of analysis on label

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

pH 6 to 8
Melting Point -61°C
Color Colorless
Decomposition Information >350°C
Density 0.945g/cm³
Quantity 1 L
Flash Point 58°C (136.4°F)
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.005% max.
Assay Percent Range ≥99.8 %
Titratable Acid 0.0005mEq/g max.
Titratable Base 0.003mEq/g max.
Vapor Pressure 4.9mbar at 20°C
Viscosity 0.8 mPa/s at 20°C
UN Number 2265
Water 0.15% max.
Boiling Point 153°C
Formula Weight 73.09
Percent Purity ≥99.8%
Grade ACS
Chemical Name or Material N, N-Dimethylformamide
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. May cause methemoglobinemia. Irritating to eyes and skin. Lachrymator (substance which increases the flow of tears). May cause central nervous system effects. May cause adverse liver effects. May cause adverse kidney effects. May cause harm to the unborn child. Use personal protective equipment. Keep away from open flames
DOTInformation DOT Class 3, : Flammable Liquid