missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, B&J Brand™, for HPLC, GC and spectrophotometry, >99.9%, Honeywell Burdick & Jackson

Catalog No. p-7111736
Change view
Click to view available options
Quantity:
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001009 6 x 1 L Glass Bottle
6001008 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001009 Supplier Honeywell Chemicals Supplier No. 0761L
Only null left

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Absorbance 0.3 AU max. at 275 nm (UV), 0.05 AU max. at 300 nm (UV), 0.005 AU max. at 350 nm (UV), 0.005 AU max. at 400 nm (UV)
CAS Max % 100.0000%
Density 0.949 g/cm3
Boiling Point 153°C
Quantity 6 x 1 L
Flash Point 58°C
Packaging Glass Bottle
UN Number UN2265
Beilstein 605365
Water 0.03% max.
Solubility Information Miscible in Water at 20°C
Formula Weight 73.10
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material N,N-Dimethylformamide, B&J Brand™, for HPLC, GC and spectrophotometry, >99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: N,N-Dimethylformamide