missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, Anhydrous, 99.8% min., DriSolv™, MilliporeSigma™

Catalog No. MDX17276
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MDX17276 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MDX17276 Supplier Technidata Supplier No. US1DX1727/6

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Color Colorless
Quantity 1 L
Physical Form Liquid
Assay Percent Range 99.8% min.
Formula Weight 73.09
Chemical Name or Material N, N-Dimethylformamide, Anhydrous