missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.8%, Solstice

Catalog No. 6001010
Change view
Click to view available options
Quantity:
19 L
Packaging:
Metal Can
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001010 19 L Metal Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6001010 Supplier Honeywell Inc Supplier No. LP07619

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

CAS Max % 100.0000%
Density 0.949 g/cm3
Quantity 19 L
Flash Point 58°C
Packaging Metal Can
UN Number UN2265
Water 0.15% max.
Beilstein 605365
Boiling Point 153°C
Solubility Information Miscible in Water at 20°C
Formula Weight 73.10
Percent Purity >99.8%
Grade ACS
Assay by GC Analysis
Chemical Name or Material N,N-Dimethylformamide, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.8%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: N,N-Dimethylformamide