missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, ACS, 99.8+%

Catalog No. p-7082648
Change view
Click to view available options
Quantity:
4 L
4 x 4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39117K7 4 L
AA39117M6 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA39117K7 Supplier Thermo Scientific Chemicals Supplier No. 039117K7
Only null left

A solvent for chromogenic substrates | CAS: 68-12-2 | C3H7NO | 73.10 g/mol

N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

pH 7
Melting Point -61°C
Density 0.944
Boiling Point 153°C
Quantity 4 L
Flash Point 57°C (135°F)
Odor Amine-like
Refractive Index 1.431
UN Number UN2265
Beilstein 605365
Merck Index 14,3243
Solubility Information Miscible with water and most organic solvents.
Formula Weight 73.09
Percent Purity ≥99.8%
Grade ACS
Chemical Name or Material N,N-Dimethylformamide
Show More Show Less
Hazard Category H226-H312+H332-H319-H335-H336-H350-H360D
Hazard Statement GHS H Statement
H360-H226-H312-H332-H319;
May damage fertility or the unborn child.
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P312-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: N,N-DIMETHYLFORMAMIDE
EINECSNumber 200-679-5
RTECSNumber LQ2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only