missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, ACS, 99.8%, Spectrum™ Chemical

Catalog No. p-7165456
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
20 L
55 gal.
Packaging:
Amber Glass Bottle
Drum
Steel Can
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611875 100 mL Amber Glass Bottle
18611878 500 mL Amber Glass Bottle
18611876 1 L Amber Glass Bottle
18611877 4 L Amber Glass Bottle
18611873 20 L Steel Can
18611874 55 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18611875 Supplier Spectrum Chemical Mfg Cor Supplier No. D1030100ML
Only null left

Chemical Identifiers

CAS 68-12-2
Molecular Weight (g/mol) 73.09

Specifications

CAS Max % 1
Boiling Point 153°C
Quantity 100 mL
Identification To Pass Test
Packaging Amber Glass Bottle
Residue after Evaporation 5e-05
Titratable Acid 0.0005 meq/g
Titratable Base 0.003 meq/g
Water 0.0015
Percent Purity 0.998
Grade Reagent
Chemical Name or Material N,N-Dimethylformamide
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.