missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylformamide, ACS, 99.8%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 55 gal. |
| Identification | To Pass Test |
| Packaging | Drum |
| Residue after Evaporation | 5e-05 |
| Titratable Acid | 0.0005 meq/g |
| Titratable Base | 0.003 meq/g |
| Water | 0.0015 |
| Boiling Point | 153°C |
| Percent Purity | 0.998 |
| Show More |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.