missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, 99%

CAS: 68-12-2 | C3H7NO | 73.10 g/mol

Supplier:  Thermo Scientific Chemicals A13547AK

 View more versions of this product

Catalog No. AAA13547AK


Only null left
Explore available promotions

Description

Description

N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films.

Solubility
Miscible with water and most organic solvents.

Notes
Reaction with sodium hydride gives exothermic decomposition at low temperatures. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Specifications

N,N-Dimethylformamide
-61°C
7
57°C (135°F)
C3H7NO
MFCD00003284
UN2265
14,3243
Miscible with water and most organic solvents.
CN(C)C=O
73.10
CHEBI:17741
99%
68-12-2
0.944
153°C
Amine-like
1.431
250 mL
605365
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
ZMXDDKWLCZADIW-UHFFFAOYSA-N
N,N-dimethylformamide
6228
73.09

Chemical Identifiers

68-12-2
73.10
ZMXDDKWLCZADIW-UHFFFAOYSA-N
6228
N,N-dimethylformamide
C3H7NO
MFCD00003284
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
CHEBI:17741
CN(C)C=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360-H226-H312-H332-H319
May damage fertility or the unborn child.
Flammable liquid and vapour.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P312-P363-P370+P378q-P501c

H226-H312+H332-H319-H335-H336-H350-H360D

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: N,N-DIMETHYLFORMAMIDE

EINECSNumber : 200-679-5

RTECSNumber : LQ2100000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only