Learn More
N,N-Dimethylformamide, 99.9%, for biochemistry, AcroSeal™
CAS: 68-12-2 | C3H7NO | 73.10 g/mol
$263.17 - $263.17
Chemical Identifiers
| CAS | 68-12-2 |
|---|---|
| Molecular Formula | C3H7NO |
| Molecular Weight (g/mol) | 73.10 |
| MDL Number | MFCD00003284 |
| InChI Key | ZMXDDKWLCZADIW-UHFFFAOYSA-N |
| Synonym | dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa |
| PubChem CID | 6228 |
| ChEBI | CHEBI:17741 |
| IUPAC Name | N,N-dimethylformamide |
| SMILES | CN(C)C=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC447090010
|
Thermo Scientific Chemicals
447090010 |
1 L | AcroSeal™ Glass bottle |
Each for $263.17
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 68-12-2 | |
| 73.10 | |
| ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
| 6228 | |
| N,N-dimethylformamide |
| C3H7NO | |
| MFCD00003284 | |
| dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
| CHEBI:17741 | |
| CN(C)C=O |
Specifications
| 68-12-2 | |
| 0.9450g/mL | |
| 58°C | |
| AcroSeal™ Glass bottle | |
| 1.4290 to 1.4320 | |
| MFCD00003284 | |
| 0.945 | |
| 0.94 | |
| Solubility in water: soluble. Other solubilities: miscible with most organic solvents | |
| CN(C)C=O | |
| 73.10 | |
| 0.8 mPa.s (20°C) | |
| 73.09 | |
| Biochemistry |
| -61.0°C | |
| 153.0°C | |
| 99.85% min. (GC) | |
| C3H7NO | |
| HCON(CH3)2 | |
| 1 L | |
| 15,3267 | |
| dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
| ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
| N,N-dimethylformamide | |
| 6228 | |
| CHEBI:17741 | |
| 99.9% | |
| N,N-Dimethylformamide |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May damage the unborn child.
Flammable liquid and vapour.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wea
GHS Signal Word: Danger
EINECSNumber : 200-679-5
RUO – Research Use Only