missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylformamide, 99.8%, for spectroscopy
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
Specifications
Specifications
| Alkalinity | 0.001 meq/g max. |
| Acidity | 0.001 meq/g max. |
| Absorbance | (1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.01 max. at 340nm, 0.05 max. at 310nm, 0.1 max. at 295nm, 0.3 max. at 275nm, 1 max. at 270nm |
| CAS Min % | 99.75 |
| CAS Max % | 100.0 |
| Melting Point | -61.0°C |
| Density | 0.9450g/mL |
| Boiling Point | 153.0°C |
| Flash Point | 58°C |
| Assay Percent Range | 99.75% min. (GC) |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes serious eye irritation. Harmful in contact with skin. Harmful if inhaled. May damage the unborn child. Flammable liquid and vapour. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Wea |
| EINECSNumber | 200-679-5 |
| RTECSNumber | LQ2100000 |
| TSCA | TSCA |
RUO – Research Use Only