missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, 99.8% min., ACS, MilliporeSigma™

Catalog No. MDX173020
Change view
Click to view available options
Quantity:
4 L
200 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MDX173020 200 L
MDX17301 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MDX173020 Supplier Technidata Supplier No. DX173020

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.1
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Color Colorless
Quantity 200 L
Physical Form Liquid
Assay Percent Range 99.8% min.
Formula Weight 73.09
Chemical Name or Material N, N-Dimethylformamide, For ACS analysis