missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Diméthylformamide, 99,8%, extra sec, AcroSeal™, Thermo Scientific Chemicals
Description
Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.
Spécifications
Spécifications
| Nom ou substance chimique | N, N-Dimethylformamide |
| Point de fusion | -61 °C |
| Renseignements sur la décomposition | >350 °C |
| Densité | 0.9450g/mL |
| Quantité | 1 L |
| Point d’éclair | 58 °C (136,4 °F) |
| Désignation | Test Réussi |
| Fourchette de pourcentages d’essai | 99.8% |
| Pression de vapeur | Pression de vapeur : 4,9 mbar à 20 °C |
| Formule linéaire | HCON(CH3)2 |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement: Causes serious eye irritation. Harmful in contact with skin. Harmful if inhaled. May damage the unborn child. Flammable liquid and vapour. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 200-679-5 |
| RTECSNumber | LQ2100000 |
| TSCA | TSCA |
RUO – Usage réservé à la recherche