missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, 99.5%, for HPLC
GREENER_CHOICE

Catalog No. AC279600010
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC279600010 1 L Glass Bottle
AC279600025 2.5 L Glass Bottle
AC279600040 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC279600010 Supplier Thermo Scientific Chemicals Supplier No. 279600010
Get 1 Extra Chemical When You Buy 2
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For HPLC | CAS: 68-12-2 | C3H7NO | 73.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Melting Point -61.0°C
Absorbance (1 cm cell vs HPLC−grade water), (1 cm cell vs HPLC−grade water) at 0nm, 0.005 max. at 350nm, 0.015 max. at 340nm, 0.02 max. at 320nm, 0.05 max. at 300nm, 0.15 max. at 280nm, 0.5 max. at 270nm
CAS Min % 99.4
CAS Max % 100.0
Acidity 0.001 meq/g max.
Alkalinity 0.001 meq/g max.
Decomposition Information >350°C
Density 0.9450g/mL
Quantity 1 L
Flash Point 58°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4290 to 1.4320
Residue after Evaporation 0.001% max.
Specific Gravity 0.945
Assay Percent Range 99.5%
Vapor Pressure Vapor Pressure: 4.9mbar at 20°C
Linear Formula HCON(CH3)2
Viscosity 0.8 mPa.s (20°C)
UN Number 2265
Water 0.05% max. (Coulometric)
Beilstein 04, 58
Fieser 01,278; 02,153; 03,115; 04,184; 05,247; 07,124; 08,189; 09,182; 11,198; 12,203; 13,124; 14,148; 16,144
Merck Index 15, 3267
Green Features ACT Ecolabel (Silver)
Boiling Point 153.0°C
Solubility Information Solubility in water: soluble. Other solubilities: miscible with most organic solvents
Formula Weight 73.09
Percent Purity 99.5%
Grade HPLC
Chemical Name or Material N,N-Dimethylformamide
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May damage the unborn child.
Flammable liquid and vapour.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-679-5
RTECSNumber LQ2100000
TSCA TSCA

RUO – Research Use Only