missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dimethylethylenediamine 97.0+%, TCI America™

Catalog No. D07205ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D07205ML 5 mL
D072025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D07205ML Supplier TCI America Supplier No. D07205ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-70-3
Molecular Formula C4H12N2
Molecular Weight (g/mol) 88.154
MDL Number MFCD00008290
InChI Key KVKFRMCSXWQSNT-UHFFFAOYSA-N
Synonym n,n'-dimethylethylenediamine, 1,2-bis methylamino ethane, n1,n2-dimethylethane-1,2-diamine, n,n'-dimethyl-1,2-ethanediamine, dimethylethylenediamine, sym-dimethylethylenediamine, 1,2-ethanediamine, n,n'-dimethyl, 2,5-diazahexane, n,n'-dimethyldiaminoethane, n,n'-dimethylethanediamine
PubChem CID 8070
IUPAC Name N,N'-dimethylethane-1,2-diamine
SMILES CNCCNC

Specifications

Color Yellow
Boiling Point 119°C
Quantity 5 mL
UN Number 2734
Formula Weight 88.15
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N′-Dimethylethylenediamine
RTECSNumber KV4250000
TSCA Yes