missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylethylenediamine, 97%

Alfa Aesar N,N-Dimethylethylenediamine, 97%, Quantity: 250g, Boiling Point: 107 deg.C, Melting Point: <-70 deg.C, Molecular Weight: 88.154, Percent Purity: 97%, Beilstein: 605279, CAS: 108-00-9, Density: 0.81, DOT Information: Hazard Class: 8; Packaging Group: I, EINECS Number: 203-541-2 | CAS: 108-00-9 | C4H12N2 | 88.154 g/mol

Supplier:  Thermo Scientific Chemicals B2018222

 View more versions of this product

Catalog No. AAB2018222

Only null left
Explore available promotions

Description

Description

It finds its application as an intermediate in the synthesis of Cefotiam. It is also used as a catalyst in the synthesis of allylic nitro compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as an intermediate in the synthesis of Cefotiam. It is also used as a catalyst in the synthesis of allylic nitro compounds.

Solubility
Fully miscible in water.

Notes
Air sensitive & hygroscopic. Store away from oxidizing agents, air and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from electrostatic charges.
Specifications

Specifications

N,N-Dimethylethylenediamine
<-70°C
107°C
C4H12N2
(CH3)2NCH2CH2NH2
100 g
605279
n,n-dimethylethylenediamine, 2-dimethylaminoethylamine, 2-aminoethyl dimethylamine, 1,2-ethanediamine, n,n-dimethyl, 2-dimethylamino ethylamine, n,n-dimethyl-1,2-ethanediamine, n1,n1-dimethylethane-1,2-diamine, n,n-dimethyl-1,2-ethylenediamine, 2-aminoethyldimethylamine, n,n-dimethylethanediamine
DILRJUIACXKSQE-UHFFFAOYSA-N
N',N'-dimethylethane-1,2-diamine
66053
97%
108-00-9
0.81
12°C (53°F)
1.427
MFCD00008175
UN2733
Air sensitive; Hygroscopic
Fully miscible in water.
CN(C)CCN
88.154
88.15

Chemical Identifiers

108-00-9
88.154
DILRJUIACXKSQE-UHFFFAOYSA-N
66053
CN(C)CCN
C4H12N2
MFCD00008175
n,n-dimethylethylenediamine, 2-dimethylaminoethylamine, 2-aminoethyl dimethylamine, 1,2-ethanediamine, n,n-dimethyl, 2-dimethylamino ethylamine, n,n-dimethyl-1,2-ethanediamine, n1,n1-dimethylethane-1,2-diamine, n,n-dimethyl-1,2-ethylenediamine, 2-aminoethyldimethylamine, n,n-dimethylethanediamine
N',N'-dimethylethane-1,2-diamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H302-H312
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P210-P233-P240-P241-P242-P243-P260-P264b-P270-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P363-P372-P374-P380-P501c

H225-H302+H312-H314

DOTInformation : Hazard Class: 8; Packaging Group: I

EINECSNumber : 203-541-2

RTECSNumber : KH8608250

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only