Learn More
N,N'-Dimethylethylenediamine, 95%
CAS: 110-70-3 | C4H12N2 | 88.154 g/mol
Supplier: Thermo Scientific Chemicals L0220414
Description
N,N'-Dimethylethylenediamine is used in DNA binding effect. It is used to enhance the adsorption of carbon dioxide. It acts as a ligand and form coordination complex such as dinitrato(N,N'-dimethyl-1,2-ethanediamine)copper(II) and dichloro(1,4-bis-(diphenyl phosphino)butane)-(1,2-ethylenediamine)ruthenium(II).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N′-Dimethylethylenediamine is used in DNA binding effect. It is used to enhance the adsorption of carbon dioxide. It acts as a ligand and form coordination complex such as dinitrato(N,N′-dimethyl-1,2-ethanediamine)copper(II) and dichloro(1,4-bis-(diphenyl phosphino)butane)-(1,2-ethylenediamine)ruthenium(II).
Solubility
Miscible with chloroform and dichloromethane.
Notes
Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
Specifications
| N,N'-Dimethylethylenediamine | |
| 0.818 | |
| 28°C (82°F) | |
| C4H12N2 | |
| CH3NHCH2CH2NHCH3 | |
| 25 g | |
| 878142 | |
| n,n'-dimethylethylenediamine, 1,2-bis methylamino ethane, n1,n2-dimethylethane-1,2-diamine, n,n'-dimethyl-1,2-ethanediamine, dimethylethylenediamine, sym-dimethylethylenediamine, 1,2-ethanediamine, n,n'-dimethyl, 2,5-diazahexane, n,n'-dimethyldiaminoethane, n,n'-dimethylethanediamine | |
| KVKFRMCSXWQSNT-UHFFFAOYSA-N | |
| N,N'-dimethylethane-1,2-diamine | |
| 8070 | |
| 95% |
| 110-70-3 | |
| 118°C to 119°C | |
| Amine-like | |
| 1.43 | |
| MFCD00008290 | |
| UN2734 | |
| Air Sensitive | |
| Miscible with chloroform and dichloromethane. | |
| CNCCNC | |
| 88.154 | |
| 88.15 |
Chemical Identifiers
| 110-70-3 | |
| 88.154 | |
| KVKFRMCSXWQSNT-UHFFFAOYSA-N | |
| 8070 | |
| CNCCNC |
| C4H12N2 | |
| MFCD00008290 | |
| n,n'-dimethylethylenediamine, 1,2-bis methylamino ethane, n1,n2-dimethylethane-1,2-diamine, n,n'-dimethyl-1,2-ethanediamine, dimethylethylenediamine, sym-dimethylethylenediamine, 1,2-ethanediamine, n,n'-dimethyl, 2,5-diazahexane, n,n'-dimethyldiaminoethane, n,n'-dimethylethanediamine | |
| N,N'-dimethylethane-1,2-diamine |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
EINECSNumber : 203-793-3
RTECSNumber : KV4250000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only