missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylethylamine, 99%, Thermo Scientific™

Catalog No. AC219290010
Change view
Click to view available options
1L; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC219290010 1L; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC219290010 Supplier Thermo Scientific Chemicals Supplier No. 219290010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 598-56-1
CAS Min % 98.5
CAS Max % 100.0
Density 0.6750g/mL
Boiling Point 36°C
Flash Point -36°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C4H11N
Refractive Index 1.3715 to 1.3735
Linear Formula C2H5N(CH3)2
Quantity 1L
MDL Number MFCD00009039
Beilstein 04, 94
Specific Gravity 0.675
Synonym n,n-dimethylethylamine, dimethylethylamine, ethanamine, n,n-dimethyl, ethyldimethylamine, n-ethyldimethylamine, ethylamine, n,n-dimethyl, methanamine, n-ethyl-n-methyl, unii-9n5384xvem, c13-15-alkyldimethylamines, amines, c13-15-alkyldimethyl
Solubility Information Solubility in water: soluble. Other solubilities: miscible with most common organic solvents
InChI Key DAZXVJBJRMWXJP-UHFFFAOYSA-N
SMILES CCN(C)C
IUPAC Name N,N-dimethylethanamine
Molecular Weight (g/mol) 73.13
PubChem CID 11723
Formula Weight 73.13
Percent Purity 99%
Chemical Name or Material N, N-Dimethylethylamine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapour.
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth
EINECSNumber 209-940-8
TSCA TSCA

RUO – Research Use Only