missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyldodecylamine 96.0+%, TCI America™

Catalog No. p-7125975
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
D000225ML 25 mL
D0002500ML 500 mL
2 options

Catalog No. D000225ML

Supplier: TCI America D000225ML

Only null left

Chemical Identifiers

CAS 112-18-5
Molecular Formula C14H31N
Molecular Weight (g/mol) 213.409
MDL Number MFCD00008970
InChI Key YWFWDNVOPHGWMX-UHFFFAOYSA-N
Synonym n,n-dimethyldodecylamine, lauryldimethylamine, dodecyldimethylamine, antioxidant dda, dimethyl lauramine, n,n-dimethyl-n-dodecylamine, 1-dodecanamine, n,n-dimethyl, n,n-dimethyllaurylamine, dda antioxidant, barlene 125
PubChem CID 8168
IUPAC Name N,N-dimethyldodecan-1-amine
SMILES CCCCCCCCCCCCN(C)C

Specifications

Melting Point -20°C
Color Yellow
Boiling Point 149°C
Quantity 25 mL
UN Number 2735
Formula Weight 213.41
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Dimethyldodecylamine
EINECSNumber (2)-0176
RTECSNumber JR6600000
TSCA Yes