missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyldodecylamine, 95%, pract.

Catalog No. AC408292500
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC408292500 250 g Glass bottle
AC408290010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC408292500 Supplier Thermo Scientific Chemicals Supplier No. 408292500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 112-18-5 | C14H31N | 213.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112-18-5
Molecular Formula C14H31N
Molecular Weight (g/mol) 213.41
MDL Number MFCD00008970
InChI Key YWFWDNVOPHGWMX-UHFFFAOYSA-N
Synonym n,n-dimethyldodecylamine, lauryldimethylamine, dodecyldimethylamine, antioxidant dda, dimethyl lauramine, n,n-dimethyl-n-dodecylamine, 1-dodecanamine, n,n-dimethyl, n,n-dimethyllaurylamine, dda antioxidant, barlene 125
PubChem CID 8168
IUPAC Name N,N-dimethyldodecan-1-amine
SMILES CCCCCCCCCCCCN(C)C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point -20.0°C
Density 0.7870g/mL
Boiling Point 271.0°C
Flash Point 118°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Dodecyl) (GC)
Packaging Glass bottle
Refractive Index 1.4365 to 1.4385
Linear Formula CH3(CH2)11N(CH3)2
Quantity 250 g
Specific Gravity 0.787
Solubility Information Solubility in water: negligible
Viscosity 1.8 mPa.s (100°C)
Formula Weight 213.41
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material N, N-Dimethyldodecylamine, pract., 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-943-8
RTECSNumber JR6600000
TSCA TSCA

RUO – Research Use Only