missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyldodecylamine, 90+%

Catalog No. AAB2414530
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2414530 250 g
AAB2414518 50 g
AAB241450B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2414530 Supplier Thermo Scientific Chemicals Supplier No. B2414530
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 112-18-5 | C14H31N | 213.409 g/mol

N,N-Dimethyldodecylamine is used as a as a capping agent during the synthesis of Au(core) -Pd(shell) bimetallic nanoparticles in toluene. It is used in the synthesis of N-alkyl-N,N-dimethyl-1-ammonio-2-hydroxy-3-propane sulfonate. Also used in the preparation of novel bis-quaternary ammonium salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyldodecylamine is used as a as a capping agent during the synthesis of Au(core) -Pd(shell) bimetallic nanoparticles in toluene. It is used in the synthesis of N-alkyl-N,N-dimethyl-1-ammonio-2-hydroxy-3-propane sulfonate. Also used in the preparation of novel bis-quaternary ammonium salt.

Solubility
Slightly soluble in water.

Notes
Air & moisture sensitive. Materials to avoid is copper, oxidizing agents, strong acids.

Chemical Identifiers

CAS 112-18-5
Molecular Formula C14H31N
Molecular Weight (g/mol) 213.409
MDL Number MFCD00008970
InChI Key YWFWDNVOPHGWMX-UHFFFAOYSA-N
Synonym n,n-dimethyldodecylamine, lauryldimethylamine, dodecyldimethylamine, antioxidant dda, dimethyl lauramine, n,n-dimethyl-n-dodecylamine, 1-dodecanamine, n,n-dimethyl, n,n-dimethyllaurylamine, dda antioxidant, barlene 125
PubChem CID 8168
IUPAC Name N,N-dimethyldodecan-1-amine
SMILES CCCCCCCCCCCCN(C)C

Specifications

Melting Point -20°C
Density 0.787
Boiling Point 110°C to 112°C (3 mmHg)
Flash Point 118°C (244°F)
Refractive Index 1.4375
Quantity 250 g
UN Number UN2735
Sensitivity Air and moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 213.41
Percent Purity ≥90%
Chemical Name or Material N,N-Dimethyldodecylamine
Show More Show Less
Hazard Category H302-H314
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 203-943-8
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only