missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethyldecylamine 95.0+%, TCI America™
$39.14 - $70.84
Chemical Identifiers
| CAS | 1120-24-7 |
|---|---|
| Molecular Formula | C12H27N |
| Molecular Weight (g/mol) | 185.36 |
| MDL Number | MFCD00015060 |
| InChI Key | YWWNNLPSZSEZNZ-UHFFFAOYSA-N |
| Synonym | 1-(Dimethylamino)decane |
| PubChem CID | 70711 |
| IUPAC Name | decyldimethylamine |
| SMILES | CCCCCCCCCCN(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D228825ML
|
TCI America
D228825ML |
25 mL |
Each for $39.14
|
|
|||||
|
D2288500ML
|
TCI America
D2288500ML |
500 mL |
Each for $70.84
|
|
|||||
Chemical Identifiers
| 1120-24-7 | |
| 185.36 | |
| YWWNNLPSZSEZNZ-UHFFFAOYSA-N | |
| 70711 | |
| CCCCCCCCCCN(C)C |
| C12H27N | |
| MFCD00015060 | |
| 1-(Dimethylamino)decane | |
| decyldimethylamine |
Specifications
| 1120-24-7 | |
| 235°C | |
| MFCD00015060 | |
| 2735 | |
| YWWNNLPSZSEZNZ-UHFFFAOYSA-N | |
| decyldimethylamine | |
| 70711 | |
| ≥95.0% (GC,T) | |
| N,N-Dimethyldecylamine |
| Colorless | |
| C12H27N | |
| 25 mL | |
| 1-(Dimethylamino)decane | |
| CCCCCCCCCCN(C)C | |
| 185.36 | |
| 185.36 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0176
TSCA : Yes