missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyldecanamide 98.0+%, TCI America™

Catalog No. D444825ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D444825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D444825ML Supplier TCI America Supplier No. D444825ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14433-76-2
Molecular Formula C12H25NO
Molecular Weight (g/mol) 199.338
MDL Number MFCD00043725
InChI Key HNXNKTMIVROLTK-UHFFFAOYSA-N
Synonym decanamide, n,n-dimethyl, n,n-dimethylcapramide, n,n-dimethylcapylamide, n,n-dimethyldecanoamide, unii-o29y6x2jez, n,n-dimethyldecan-1-amide, nn-dimethyldecanamide, n,n-dimethyl decanamide, o29y6x2jez, dimethyl capramide
PubChem CID 26690
IUPAC Name N,N-dimethyldecanamide
SMILES CCCCCCCCCC(=O)N(C)C

Specifications

Color Yellow
Boiling Point 110°C
Quantity 25 mL
Formula Weight 199.34
Percent Purity ≥98.0% (GC)
Grade Reagent
Physical Form Liquid
Chemical Name or Material N,N-Dimethyldecanamide
EINECSNumber (2)-0825
RTECSNumber HD6350000
TSCA Yes