missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylcyclohexylamine, 98+%

Catalog No. p-7046481
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL14521AC 25 mL
AAL14521AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL14521AC Supplier Thermo Scientific Chemicals Supplier No. L14521AC
Only null left

CAS: 98-94-2 | C8H17N | 127.231 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98-94-2
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.231
MDL Number MFCD00003844
InChI Key SVYKKECYCPFKGB-UHFFFAOYSA-N
Synonym n,n-dimethylcyclohexylamine, cyclohexyldimethylamine, dimethylcyclohexylamine, n-cyclohexyldimethylamine, polycat 8, cyclohexanamine, n,n-dimethyl, dimethylamino cyclohexane, n,n-dimethylaminocyclohexane, n,n-dimethyl-n-cyclohexylamine, cyclohexylamine, n,n-dimethyl
PubChem CID 7415
ChEBI CHEBI:59022
IUPAC Name N,N-dimethylcyclohexanamine
SMILES CN(C)C1CCCCC1

Specifications

Melting Point -60°C
Density 0.849
Boiling Point 160°C to 161°C
Flash Point 39°C (102°F)
Refractive Index 1.454
Quantity 25 mL
UN Number UN2264
Beilstein 1919922
Formula Weight 127.23
Percent Purity ≥98%
Chemical Name or Material N,N-Dimethylcyclohexylamine
Show More Show Less
Hazard Category H226-H301+H311+H331-H314
Hazard Statement GHS H Statement
H310-H330-H314-H318-H226-H302
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: N,N-DIMETHYLCYCLOHEXYLAMINE
EINECSNumber 202-715-5
RTECSNumber GX1198000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only