missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylanilinium Tetrakis(pentafluorophenyl)borate 98.0+%, TCI America™

Catalog No. p-7128257
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4404200MG 200 mg
D44041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4404200MG Supplier TCI America Supplier No. D4404200MG
Only null left

Chemical Identifiers

CAS 118612-00-3
Molecular Formula C32H12BF20N
Molecular Weight (g/mol) 801.233
MDL Number MFCD01074420
InChI Key BRHZQNMGSKUUMN-UHFFFAOYSA-O
Synonym dimethylanilinium tetrakis pentafluorophenyl borate, unii-h8r86l92nd, n,n-dimethylanilinium tetrakis pentafluorophenyl borate, n,n-dimethylbenzenaminium tetrakis perfluorophenyl borate, n,n-dimethylanilinium tetra pentafluorophenyl borate, benzene, pentafluoro-, boron complex, n,n-dimethylanilinium tetrakis pentafluorophenyl borate 1-, benzenamine, n,n-dimethyl-, tetrakis pentafluorophenyl borate 1-, dimethyl phenyl ammonium tetrakis 2,3,4,5,6-pentafluorophenyl borate, borate 1-, tetrakis pentafluorophenyl-, hydrogen, compd. with n,n-dimethylbenzenamine 1:1:1
PubChem CID 10996402
IUPAC Name dimethyl(phenyl)azanium;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
SMILES [B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.C[NH+](C)C1=CC=CC=C1

Specifications

Melting Point 227°C
Color White
Quantity 200 mg
Formula Weight 801.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Dimethylanilinium Tetrakis(pentafluorophenyl)borate
EINECSNumber (3)-4390
TSCA No