missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylaminomethylferrocene 97.0+%, TCI America™

Catalog No. D12715G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12715G 5 g
D127125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D12715G Supplier TCI America Supplier No. D12715G

Chemical Identifiers

CAS 1271-86-9
Molecular Formula C13H17FeN
Molecular Weight (g/mol) 243.13
MDL Number MFCD00001433
InChI Key JJJSTEANRWLZBH-UHFFFAOYNA-N
Synonym Ferrocenylmethyldimethylamine
IUPAC Name (Dimethylaminomethyl)ferrocene
SMILES [Fe].c1cccc1.CN(C)Cc1cccc1

Specifications

Color Brown
Quantity 5 g
Formula Weight 243.13
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Dimethylaminomethylferrocene