missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacrylamide (stabilized with MEHQ) 99.0+%, TCI America™

Catalog No. D109125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D109125G 25 g
D1091500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D109125G Supplier TCI America Supplier No. D109125G

Chemical Identifiers

CAS 2680-03-7
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
MDL Number MFCD00008626
InChI Key YLGYACDQVQQZSW-UHFFFAOYSA-N
Synonym n,n-dimethylacrylamide, 2-propenamide, n,n-dimethyl, acrylamide, n,n-dimethyl, acylamide, n,n-dimethyl, n,n-dimethyl-acrylamide, n,n-dimethyl-2-propenamide, dimethylamid kyseliny akrylove, nn-dimethylacrylamide, unii-as46jk7q6i, propenamide, n,n-dimethyl
PubChem CID 17587
IUPAC Name N,N-dimethylprop-2-enamide
SMILES CN(C)C(=O)C=C

Specifications

Color Yellow
Boiling Point 80°C
Quantity 25 g
UN Number 2810
Formula Weight 99.13
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Dimethylacrylamide (stabilized with MEHQ)
EINECSNumber (2)-1017
RTECSNumber AU3230000
TSCA Yes