missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetoacetamide 97.0+%, TCI America™

Catalog No. D064225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D064225ML 25 mL
D0642500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D064225ML Supplier TCI America Supplier No. D064225ML
Only null left

Chemical Identifiers

CAS 2044-64-6
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD00038243
InChI Key YPEWWOUWRRQBAX-UHFFFAOYSA-N
Synonym n,n-dimethylacetoacetamide, butanamide, n,n-dimethyl-3-oxo, n,n-dimethyl-3-oxobutyramide, acetylacetamide, n,n-dimethyl, unii-7e2l0oaa4k, acetoacetamide, n,n-dimethyl, dimethylamid kyseliny acetoctove, dimethyl acetoacetamide, n,n-dimethyl acetoacetamide, nn-dimethylacetoacetamide
PubChem CID 16278
IUPAC Name N,N-dimethyl-3-oxobutanamide
SMILES CC(=O)CC(=O)N(C)C

Specifications

Color Yellow
Boiling Point 220°C
Quantity 25 mL
Formula Weight 129.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Dimethylacetoacetamide
EINECSNumber (2)-2767
RTECSNumber AK4035000
TSCA Yes