Learn More
N,N-Dimethylacetamide, HPLC Grade, 99.5+%
CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Supplier: Thermo Scientific Chemicals 022916M1
Description
N,N-Dimethylacetamide is used in all analytical laboratories as a major component of mobile phase in High Performance Liquid Chromatography (HPLC) and in Liquid Chromatography - Mass Spectrometry (LC-MS).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| N, N-Dimethylacetamide | |
| 127-19-5 | |
| Colorless liquid | |
| 165°C to 166°C | |
| 70°C (158°F) | |
| 1.4385 | |
| C4H9NO | |
| 3.3 hPa (3mm Hg) at 20°C | |
| dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine | |
| FXHOOIRPVKKKFG-UHFFFAOYSA-N | |
| N,N-dimethylacetamide | |
| 31374 | |
| 87.12 |
| -20°C | |
| Colorless | |
| 0.94 | |
| 4 x 1 L | |
| Liquid | |
| >99.5% | |
| MFCD00008686 | |
| 1737614 | |
| Miscible with water,benzene,alcohol,acetone,ether,esters and ketones. | |
| CN(C)C(C)=O | |
| 87.12 | |
| CHEBI:84254 | |
| HPLC |
Chemical Identifiers
| 127-19-5 | |
| 87.12 | |
| FXHOOIRPVKKKFG-UHFFFAOYSA-N | |
| 31374 | |
| N,N-dimethylacetamide |
| C4H9NO | |
| MFCD00008686 | |
| dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine | |
| CHEBI:84254 | |
| CN(C)C(C)=O |
Safety and Handling
P201-P202-P210-P261-P271-P281-P302+P352-P304+P340-P308+P313-P312-P363-P370+P378q-P501c
H227-H312+H332-H360D
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 204-826-4
RTECSNumber : AB7700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only