missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide, HPLC Grade, 99.5+%

Catalog No. AA22916K2
Change view
Click to view available options
Quantity:
1 L
4 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA22916K2 1 L
AA22916M1 4 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA22916K2 Supplier Thermo Scientific Chemicals Supplier No. 022916K2

CAS: 127-19-5 | C4H9NO | 87.12 g/mol

N,N-Dimethylacetamide is used in all analytical laboratories as a major component of mobile phase in High Performance Liquid Chromatography (HPLC) and in Liquid Chromatography - Mass Spectrometry (LC-MS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 127-19-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00008686
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
PubChem CID 31374
ChEBI CHEBI:84254
IUPAC Name N,N-dimethylacetamide
SMILES CN(C)C(C)=O

Specifications

Melting Point -20°C
Color Colorless
Appearance Colorless liquid
Density 0.940 g/cm3 at 20°C
Quantity 1 L
Flash Point 70°C (158°F)
Physical Form Liquid
Refractive Index 1.4385
Assay Percent Range >99.5%
Vapor Pressure 3.3 hPa (3mm Hg) at 20°C
Beilstein 1737614
Boiling Point 165°C to 166°C
Solubility Information Miscible with water,benzene,alcohol,acetone,ether,esters and ketones.
Formula Weight 87.12
Grade HPLC
Chemical Name or Material N, N-Dimethylacetamide
Show More Show Less
Hazard Category H227-H312+H332-H360D
Precautionary Statement P201-P202-P210-P261-P271-P281-P302+P352-P304+P340-P308+P313-P312-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 204-826-4
RTECSNumber AB7700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only