missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide, HPLC Grade, 99.5%, Spectrum™ Chemical

Catalog No. p-7165707
Change view
Click to view available options
Quantity:
4 L
200 L
Packaging:
Amber Glass Bottle
Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611854 4 L Amber Glass Bottle
18611855 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18611854 Supplier Spectrum Chemical Mfg Cor Supplier No. HP5624LT
Only null left

Chemical Identifiers

CAS 127-19-5
Molecular Weight (g/mol) 87.12

Specifications

CAS Max % 1
Appearance Clear, Colorless
Quantity 4 L
Packaging Amber Glass Bottle
Residue after Evaporation 5 ppm
Water 0.0003
Boiling Point 163°C to 165°C
Percent Purity 0.995
Grade HPLC
Chemical Name or Material N,N-Dimethylacetamide

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.