missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylacetamide (DMA), Reagents
Specifications
Specifications
| Quantity | 4 L |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Specific Gravity | 0.94 |
| Quantity | 4 L |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Specific Gravity | 0.94 |