missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide (DMA), Reagents

Catalog No. 01338010
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338010 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01338010 Supplier Labchem Inc Supplier No. C11961004C
Only null left

N,N-Dimethylacetamide (DMA), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

Synonym Dimethylacetamide, DMAc

Specifications

Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.94