missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide, Certified Reference Material, MilliporeSigma™ Supelco™

Pharmaceutical secondary standard for applications in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

$295.87 - $295.87

Specifications

CAS 127-19-5
Flash Point 64°C (Not applicable)
Molecular Formula C4H9NO
Linear Formula C4H9NO
Synonym N,N-Dimethylacetamide solution
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111023062
View Documents
Merck Emd Millipore
RTC00010420ML
20 mL
Each for $295.87
Only null left
 
Description

Description

Specifications

Specifications

127-19-5
C4H9NO
N,N-Dimethylacetamide solution
Certified Reference Material
64°C (Not applicable)
C4H9NO
87.12
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'