missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylacetamide, BAKER ANALYZED™ Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
$19182.00 - $19182.00
Chemical Identifiers
| CAS | 127-19-5 |
|---|---|
| Molecular Formula | C4H9NO |
| Molecular Weight (g/mol) | 87.12 |
| MDL Number | MFCD00008686 |
| InChI Key | FXHOOIRPVKKKFG-UHFFFAOYSA-N |
| Synonym | dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine |
| PubChem CID | 31374 |
| ChEBI | CHEBI:84254 |
| IUPAC Name | N,N-dimethylacetamide |
| SMILES | CN(C)C(C)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
02003429
|
Avantor J.T.Baker
J37209 |
4 L | Narrow Mouth Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 127-19-5 | |
| 87.12 | |
| FXHOOIRPVKKKFG-UHFFFAOYSA-N | |
| 31374 | |
| N,N-dimethylacetamide |
| C4H9NO | |
| MFCD00008686 | |
| dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine | |
| CHEBI:84254 | |
| CN(C)C(C)=O |
Specifications
| 127-19-5 | |
| 70.00°C | |
| 1L = 0.94kg | |
| MFCD00008686 | |
| FXHOOIRPVKKKFG-UHFFFAOYSA-N | |
| N,N-dimethylacetamide | |
| 31374 | |
| 87.12 | |
| N,N-Dimethylacetamide |
| 4 L | |
| Narrow Mouth Amber Glass Bottle | |
| C4H9NO | |
| dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine | |
| CN(C)C(C)=O | |
| 87.12 | |
| CHEBI:84254 | |
| BAKER ANALYZED™ Reagent |